Learn More
Ly6K Antibody (CL2435) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36764F
Product Code. 30287903
  • 737.00€ / 0.10mL
Description
Ly6K Monoclonal antibody specifically detects Ly6K in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly6K
Monoclonal
FITC
cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K
This antibody was developed using a recombinant protein derived from Q17RY6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2435
PBS
Mouse
Protein A purified
RUO
54742
Store at 4⪚C in the dark.
IgG2b