missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6B.2 Antibody (7/4), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-13077PE
This item is not returnable.
View return policy
Description
Ly-6B.2 Monoclonal specifically detects Ly-6B.2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Ly-6B.2 | |
| Monoclonal | |
| PE | |
| Rat | |
| Protein G purified | |
| RUO | |
| Recognizes Ly6B.2, a polymorphic 40 kDa antigen expressed by polymorphonuclear cells, but absent on resident tissue macrophages. Strains positive for 7/4:129J; AKR; C57BL/6; C57BL/10; C58; DBA/2; NZB; NZW; SJL; MFI; Swiss (PO) Strains Negative/Weak for 7/4:A2G; A/Sn; ASW; BALB/c; C3H/HEH: CBA.T6T6. | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 7/4 | |
| Flow Cytometry | |
| Cultured bone marrow cells | |
| 0.125 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction