Learn More
Ly-6A/E Rat anti-Mouse, FITC, Clone: D7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557405
Additional Details : Weight : 0.01000kg
Description
The D7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca-1 (Ly6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cells (HSC) in the bone marrow of mice with both Ly-6 haplotypes. In mice expressing the Ly-6.2 haplotype (e.g., AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129), Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes and thymic and peripheral T lymphocytes. Strains with the Ly-6.1 haplotype (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of lymphocytes from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb (anti-Ly-6A/E, Mfr. No.553333) can block binding of FITC-conjugated D7 antibody (Mfr. No.557405) to mouse splenocytes, but purified mAb D7 is unable to block binding of FITC-conjugated E13-161.7 antibody (Mfr. No.553335). Anti-Ly-6A/E (Sca-1) mAb may be used in combination with the Mouse Lineage Panel (Mfr. No.559971) to identify HSC. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: D7
Isotype: IgG2a, κ
Cross Reactivity: Mouse
Flow Cytometry
Specifications
Ly-6A/E | |
Monoclonal | |
0.5mg/mL | |
Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
D7 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IL-2-dependent mouse T-cell line CTL-L | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.