missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 310762 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
5-HT1D-preferring receptor antagonist (EC50 = 31 nM). Displays no activity on 5-HT transport.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 430.94 |
| Purity | >98% |
| Molecular Weight (g/mol) | 430.948 |
| Chemical Name or Material | LY 310762 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction