accessibility menu, dialog, popup

UserName

H3CNONOFCH3HCl

CAS RN 192927-92-7

IUPAC Name: 1-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-3,3-dimethyl-2,3-dihydro-1H-indol-2-one hydrochloride
Synonyms: 1-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-3,3-dimethylindol-2-one hydrochloride
Molecular Weight (g/mol): 430.95
Molecular Formula: C24H28ClFN2O2
InChi Key: BOCLFQZPFYNVFD-UHFFFAOYSA-N
SMILES: Cl.CC1(C)C(=O)N(CCN2CCC(CC2)C(=O)C2=CC=C(F)C=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results