missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR beta/NR1H2 Mouse anti-Human, Clone: LXRB/2731, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | LXR beta/NR1H2 |
|---|---|
| Clone | LXRB/2731 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Protein Array |
| Applications | ELISA, Peptide Array |
Description
LXR beta/NR1H2 Monoclonal specifically detects LXR beta/NR1H2 in Human samples. It is validated for ELISA, Protein Array.Specifications
| LXR beta/NR1H2 | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| Liver X receptor beta, LXR-b, NER-I, NERliver X receptor-beta, nuclear orphan receptor LXR-beta, Nuclear receptor NER, Nuclear receptor subfamily 1 group H member 2, nuclear receptor subfamily 1, group H, member 2, oxysterols receptor LXR-beta, RIP15, steroid hormone-nuclear receptor NER, Ubiquitously-expressed nuclear receptorLXRB, UNRLX receptor beta | |
| Recombinant full-length human LXR beta/NR1H2 protein | |
| Primary | |
| Store at 4°C. |
| LXRB/2731 | |
| ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7376 | |
| IgG2c κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title