Learn More
Lubricant; Multi-Parameter Certified Reference Material, UKAS ISO 17034 / ISO 17025 Certified, 500 mL, Paragon Scientific™
Product Code. 16090050
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 16090050
Brand: Paragon Scientific CRMUMPLU
Multi-Parameter Certified Reference Materials are premium level, dual certified reference materials certified in accordance with both ISO 17025 and ISO 17034 under our UKAS accreditation. Fully dual certified in accordance with ISO 17025 and ISO 17034 under our accreditation by UKAS.
  • Certified for Viscosity, Density, Flash Point & Pour Point using ASTM / IP and EN test method protocols
  • Highest level of accreditation guarantee, providing the most credible certified data currently available world-wide
  • Low levels of uncertainty achieves maximum accuracy of data
  • Multiple test parameters provide a flexible and cost effective solution
  • All data generated exclusively by UKAS ISO 17025 accredited laboratories
  • Fully traceable to international standards
  • 18 month shelf life
  • 500mL inert glass packaging with tamper evident cap provides absolute assurance of sample integrity
  • Available from stock for immediate dispatch
  • Manufactured in the United Kingdom
Specifications
Quantity 500 mL
Nominal values quoted. Certified Values will vary from batch to batch.