missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSP1 Mouse anti-Human, Clone: LSP1/3042, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07254-100ug
This item is not returnable.
View return policy
Description
LSP1 Monoclonal specifically detects LSP1 in Human samples. It is validated for ELISA, Protein Array.
Specifications
| LSP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Protein Array | |
| 47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34 | |
| Recombinant fragment of human LSP1 protein (around aa174-278) (exact sequence is proprietary) | |
| 100 μg | |
| Phospho Specific | |
| 4046 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Peptide Array | |
| LSP1/3042 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction