missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRRC52 Monoclonal Antibody (1037552)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560869
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Auxiliary protein of the large-conductance, voltage and calcium-activated potassium channel (BK alpha). Modulates gating properties by producing a marked shift in the BK channel's voltage dependence of activation in the hyperpolarizing direction, and in the absence of calcium. KCNU1 channel auxiliary protein. May modulate KCNU1 gating properties. [UniProt].
Specifications
| LRRC52 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8N7C0 | |
| LRRC52 | |
| Human embryonic kidney cell HEK293-derived human LRRC52, Ser24-Asp244. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 1037552 | |
| Unconjugated | |
| LRRC52 | |
| 4930413P14Rik; BK channel auxiliary gamma subunit LRRC52; BK channel auxilliary gamma subunit LRRC52; leucine rich repeat containing 52; leucine-rich repeat-containing protein 52; LRRC52 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 440699 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction