missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRPPRC Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33512-100ul
This item is not returnable.
View return policy
Description
LRPPRC Monoclonal antibody specifically detects LRPPRC in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| LRPPRC | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CLONE-23970, leucine-rich PPR-motif containing, mitochondrial, mitochondrial leucine-rich PPR motif-containing protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1014-1394 of human LRPPRC (NP_573566),, Sequence:, LWYEDEKHSLNSSSASTTEPDFQKDILIACRLNQKKGAYDIFLNAKEQNIVFNAETYSNLIKLLMSEDYFTQAMEVKAFAETHIKGFTLNDAANSRLIITQVRRDYLKEAVTTLKTVLDQQQTPSRLAVTRVIQALAMKGDVENIEVVQKMLNGLEDSIGLSKMVFINNIALAQIKNNNIDAAIENIENMLTSENKVIEPQYFGLAYLFRKVIEEQLEPAVEKISIMAERLANQFAIYKPVTDFFLQLVDAGKVDDARALLQRCGAIAEQTPILLLFLLRNSRKQGKASTVKSVLELIPELNEKEEAYNSLMKSYVSEKDVTSAKALYEHLTAKNTKLDDLFLKRYASLLKYAGEPVPFIEPPESFEFYAQQLRKLRENSS | |
| 100 μL | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 10128 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction