missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRPAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | LRPAP |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 0.2-1 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638165
|
Novus Biologicals
NBP2-99704-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620966
|
Novus Biologicals
NBP2-99704-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRPAP Polyclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| LRPAP | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Signaling | |
| 0.2 um filtered solution in PBS | |
| 4043 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 0.2-1 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
| Produced in rabbits immunized with purified, recombinant Human LRPAP (Accession#: NP_002328.1; Tyr 35-Leu 357) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title