missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP4, Goat anti-Human, Polyclonal Antibody, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB18700.100ug
This item is not returnable.
View return policy
Description
Sequence: C-SNPSYRTSTQEVKSpecifications
| LRP4 | |
| Polyclonal | |
| Unconjugated | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| KIAA0816/LRP10/MEGF7 | |
| Goat | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| low density lipoprotein receptor-related protein 4 | |
| LRP4 | |
| A synthetic peptide corresponding to amino acids at internal region of human LRP4. | |
| RUO | |
| 4038 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction