missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP-1 Antibody (1C8G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52506-0.025mg
This item is not returnable.
View return policy
Description
LRP-1 Monoclonal specifically detects LRP-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Low Density LRP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| A2MRTGFBR5, Alpha-2-macroglobulin receptorMGC88725, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, FLJ16451, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, type V tgf-beta receptor | |
| Mouse | |
| 504 kDa | |
| 0.025 mg | |
| Phospho Specific | |
| 4035 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 1C8G6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| LRP1 | |
| Purified recombinant fragment of human LRP1 (AA: 20-155) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction