missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRIG3 Monoclonal Antibody (814834)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561316
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
May play a role in craniofacial and inner ear morphogenesis during embryonic development. May act within the otic vesicle epithelium to control formation of the lateral semicircular canal in the inner ear, possibly by restricting the expression of NTN1.
Specifications
| LRIG3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UXM1 | |
| LRIG3 | |
| Mouse myeloma cell line NS0-derived recombinant human LRIG3, Asp28-Thr807. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| 814834 | |
| Unconjugated | |
| LRIG3 | |
| leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 121227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction