Learn More
LRCH1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-44281G
Product Code. 18720893
  • 633.00€ / 0.10mL
Description
LRCH1 Polyclonal antibody specifically detects LRCH1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LRCH1
Polyclonal
Western Blot
Purified
Western Blot
DyLight 488
The epitope recognized by this antibody maps to a region between residue 325 to 375 of human Leucine-rich repeat and calponin homology domain-containing protein 1 using the numbering given in entry Q9Y2L9.3 (GeneID 23143).