Description
- Safe filling due to wide-necked opening 39 mm neck diameter
- Permanent print according to GHS with substance name in German, English, French and Spanish, GHS pictograms and CAS number
- LDPE yellow transparent with light protection
- Contents 500 ml
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Transparent Yellow |
| Material | LDPE |
| Quantity | 1 Ea. |
| Closure Included | Yes |
| Product Type | Safety Wash Bottle |
| Chemical Label | Unlabeled |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |