missing translation for 'onlineSavingsMsg'
Learn More

Low Temperature Viscosity Standard, JF1-H, Paragon Scientific™

Product Code. 16040200
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 16040200

Brand: Paragon Scientific JF1H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL, available also in 5L. Please contact sales@paragon-sci.com for other volumes
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Specifications

    Quantity 500 mL
    compliance-icons compliance-icons compliance-icons compliance-icons
    Product Identifier
    • 500ML JF1-H Low Temperature Viscosity Standard
    Signal Word
    • Danger
    Hazard Category
    • LONG-TERM AQUATIC HAZARD Chronic 2
    • Aspiration toxicity Category 1
    • Flammable liquid Category 3
    • Skin corrosion/irritation Category 2
    • Specific target organ toxicity Category 3
    Hazard Statement
    • H226-Flammable liquid and vapour.
    • H304-May be fatal if swallowed and enters airways.
    • H315-Causes skin irritation.
    • H336-May cause drowsiness or dizziness.
    • H411-Toxic to aquatic life with long lasting effects.
    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P273-Avoid release to the environment.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P331-Do NOT induce vomiting.
    • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
    • P391-Collect spillage.
    • P403+P235-Store in a well-ventilated place. Keep cool.
    Supplemental information
    • MIXTURE LIST-Contains : kerosine sweetened

    Nominal values quoted. Certified Values will vary from batch to batch.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.