Learn More
Low Temperature Viscosity Standard, Paragon Scientific™
Product Code. 15568874
Change view
Click to view available options
Viscosity:
151216.0 mPa.s (-40°C)
172195 mPa.s (-13°C)
5989.0 mPa.s (-40°C)
757.0 mPa.s (-40°C)
Unit Size:
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Unit Size
15568874 151216.0 mPa.s (-40°C) 500mL
15886461 757.0 mPa.s (-40°C) 500mL
15781928 5989.0 mPa.s (-40°C) 500mL
15578874 172195 mPa.s (-13°C) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15568874 Supplier Paragon Scientific Supplier No. N120B
  • ONLINE EXCLUSIVE PRICE
    112.00€
    87.63€ / 500mL
    Save 24.37€
    21% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Paragon ISO 17025 / ISO 17034 certified low temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
  • Specifications
    Chemical Name or Material N120B
    Viscosity 151216.0 mPa.s (-40°C)
    Certifications/Compliance ISO 17025, UKAS ISO 17034
    Quantity 500 mL
    Nominal values quoted