Learn More
Lenz Laborglasinstrumente™ Low-Form Weighing Bottles
Product Code. 10603971
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
45 mL
Unit Size:
Each
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10603971 30 mL Pack of 6
10508151 15 mL Each
13433589 15 mL Pack of 6
11778104 30 mL Pack of 6
15320895 45 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10603971 Supplier Lenz Laborglasinstrumente™ Supplier No. 05560513
  • ONLINE EXCLUSIVE PRICE
    127.00€
    99.95€ / Pack of 6
    Save 27.05€
    21% Off
Estimated Shipment: 22-12-2026
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Low-Form Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
  • With interchangeable NS stopper or cap
  • With fused-in size (mm) and capacity (mL)
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 30 mL
Form Low
No. per Pack 6 Pack
Height (Metric) 30 mm
Diameter (Metric) 50 mm
Closure Type Button Cover
Product Type Weighing Bottle