Learn More
Lenz Laborglasinstrumente™ Low-Form Weighing Bottles
Product Code. 10508151
Click to view available options
Capacity (Metric):
15 mL
30 mL
45 mL
Unit Size:
Each
Pack of 6
Product Code. 10508151
Brand: Lenz Laborglasinstrumente™ 05560509
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Low-Form Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
  • With interchangeable NS stopper or cap
  • With fused-in size (mm) and capacity (mL)
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 15 mL
Form Low
No. per Pack 1 Pack
Height (Metric) 30 mm
Product Type Weighing Bottle
Diameter (Metric) 35 mm
Closure Type Button Cover