Learn More
Loteprednol Etabonate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Loteprednol Etabonate
Specifications
Specifications
| Chemical Name or Material | Loteprednol Etabonate |
| CAS | 82034-46-6 |
| Analyte or Component Names | Loteprednol Etabonate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C24 H31 Cl O7 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Alrex, CDDD 5604, HGP 1, Lenoxin, Lotemax, Loteprednol etabonate, P 5604, (11beta,17alpha)-17-[(ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-Androsta-1,4-diene-17-carboxylic acid chloromethyl ester |
| InChI Formula | InChI=1S/C24H31ClO7/c1-4-30-21(29)32-24(20(28)31-13-25)10-8-17-16-6-5-14-11-15(26)7-9-22(14,2)19(16)18(27)12-23(17,24)3/h7,9,11,16-19,27H,4-6,8,10,12-13H2,1-3H3/t16-,17-,18-,19+,22-,23-,24-/m0/s1 |
| SMILES | CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)OCCl |
| IUPAC Name | chloromethyl (8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-11-hydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-17-carboxylate |
| Show More |