missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LL-37 Mouse anti-Human, Clone: B-T37, Novus Biologicals™
Mouse Monoclonal Antibody
390.00€ - 736.00€
Specifications
| Antigen | LL-37 |
|---|---|
| Clone | B-T37 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, ELISA |
| Applications | Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18342552
|
Bio-Techne
NBP3-18120-200UL |
200 μg |
390.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18380306
|
Novus Biologicals
NBP3-18120-500UL |
500 μg |
736.00€
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LL-37 Monoclonal antibody specifically detects LL-37 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| LL-37 | |
| 1 mg/ml | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS. Sterile-filtered through 0.22 um. | |
| 820 | |
| IgG1 κ | |
| Affinity purified |
| B-T37 | |
| Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
| Human antimicrobial peptide LL-37. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title