missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIV-1/Zip6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76503-0.025mg
This item is not returnable.
View return policy
Description
LIV-1/Zip6 Polyclonal specifically detects LIV-1/Zip6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LIV-1/Zip6 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25800 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| NP_001092876 | |
| SLC39A6 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP6. The immunogen is located within amino acids 430 - 480 of ZIP6. Amino Acid Squence: FKDKKKKNQKKPENDD | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction