missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liprin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58915-25ul
This item is not returnable.
View return policy
Description
Liprin alpha 1 Polyclonal specifically detects Liprin alpha 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Liprin alpha 1 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| FLJ42630, FLJ43474, LAR-interacting protein 1, LIP.1, LIP-1, LIP1FLJ41337, LIPRIN, liprin-alpha-1, Liprin-alpha1, MGC26800, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 1, PTPRF-interacting protein alpha-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8500 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| PPFIA1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HHKALDEKVRERLRVALERCSLLEEELGATHKELMILKEQNNQKKTLTDGVLDINHEQENTPSTSGKRSSDGSLSHEEDLAKVIELQEIISKQSREQSQMKERLASLSSHVTELEEDLDTAR | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido