missing translation for 'onlineSavingsMsg'
Learn More

Lipolysis Stimulated Lipoprotein Receptor Antibody, Novus Biologicals™

Product Code. 18749913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749913 0.1 mL 0.1mL
18491741 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18749913 Supplier Novus Biologicals Supplier No. NBP189631

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 11 publications

Lipolysis Stimulated Lipoprotein Receptor Polyclonal specifically detects Lipolysis Stimulated Lipoprotein Receptor in Human, Mouse, Porcine, Rhesus Macaque samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lipolysis Stimulated Lipoprotein Receptor
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated Reported in scientific publication (PMID: 32423802).
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias ILDR3, immunoglobulin-like domain containing receptor 3, lipolysis stimulated lipoprotein receptor, lipolysis-stimulated lipoprotein receptor, lipolysis-stimulated receptor, lipolysis-stimulated remnant, LISCH, LISCH protein, LISCH7, liver-specific bHLH-Zip transcription factor, MGC10659, MGC48312, MGC48503
Gene Symbols LSR
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MSEVTSLHEDDWRSRPSRGPALTPIRDEEWGGHSPRSPRGWDQEPAREQAGGGWRARRPRARSVDALDDLTPPSTAESGSRSPTSNGGRSRAYMPPRSRSRDDLYDQDDS
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51599
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Pig, Rhesus Macaque
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.