Learn More
Applied Biosystems™ Linkage Mapping Set v2.5 User Guide
Product Code. 15691762 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Manual
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15691762 1 Manual Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15691762 Supplier Applied Biosystems™ Supplier No. 4330202
In Stock
ABI PRISM™ Linkage Mapping Set Version 2.
  • For Research Use Only
  • Not for use in diagnostic procedures
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage User Guide
Product Type User Guide
Quantity 1 Manual
Shipping Condition Room Temperature