missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LINGO1 Monoclonal Antibody (332233)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561257
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors (PubMed:14966521, PubMed:15694321). Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination (PubMed:15895088). Acts in conjunction with RTN4 and RTN4R in regulating neuronal precursor cell motility during cortical development. [UniProt].
Specifications
| LINGO1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q96FE5 | |
| LINGO1 | |
| Mouse myeloma cell line NS0-derived recombinant human LINGO-1, Thr40-Thr556. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 332233 | |
| Unconjugated | |
| LINGO1 | |
| 4930471K13Rik; AV148400; LERN1; leucine rich repeat and Ig domain containing 1; leucine rich repeat neuronal 6A; leucine-rich repeat; leucine-rich repeat and immunoglobulin domain-containing protein 1; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1; leucine-rich repeat neuronal 6A; leucine-rich repeat neuronal protein 1; Leucine-rich repeat neuronal protein 6A; Lingo1; LINGO-1; LRRN6A; UNQ201; UNQ201/PRO227 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction