missing translation for 'onlineSavingsMsg'
Learn More

Lincomycin Hydrochloride Monohydrate, Mikromol™

Product Code. 17933966
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17933966

Brand: Mikromol MM0320.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lincomycin Hydrochloride Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lincomycin Hydrochloride
CAS 859-18-7
Analyte or Component Names Lincomycin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H34 N2 O6 S . Cl H
Certifications/Compliance ISO 9001
Synonym Lincomycin Hydrochloride, D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, hydrochloride (1:1), D-erythro-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)-1-thio-, monohydrochloride, trans-α- (8CI), D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[(1-methyl-4-propyl-2-pyrrolidinyl)carbonyl]amino]-1-thio-, monohydrochloride, (2S-trans)-, D-erythro-α-D-galacto-Octopyranoside, methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-, monohydrochloride (9CI), Lincomycin hydrochloride (7CI), Lincocin, Lincogap, Lincomix, NSC 70731, U 10149a, Clindamycin Hydrochloride Impurity A as Hydrochloride Monohydrate, Clindamycin Phosphate Impurity A as Hydrochloride Monohydrate, Clindamycin Impurity A as Hydrochloride Monohydrate
InChI Formula InChI=1S/C18H34N2O6S.ClH/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H/t9-,10-,11+,12-,13+,14-,15-,16-,18-;/m1./s1
SMILES Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O
IUPAC Name (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 443
Quantity 250 mg
Formula Weight 442.1904 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.