missing translation for 'onlineSavingsMsg'
Learn More

Linagliptin Acetamide, TRC

Product Code. 30137302
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137302 50 mg 50mg
30137925 25 mg 25mg
30008249 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137302 Supplier Toronto Research Chemicals Supplier No. TRCL46592050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1803079-49-3
Molecular Formula C27 H30 N8 O3
Molecular Weight (g/mol) 514.58
Synonym N-[(3R)-1-[7-(2-Butyn-1-yl)-2,3,6,7-tetrahydro-3-methyl-1-[(4-methyl-2-quinazolinyl)methyl]-2,6-dioxo-1H-purin-8-yl]-3-piperidinyl]acetamide, (R)-N-(1-(7-(But-2-ynyl)-3-methyl-1-((4-methylquinazolin-2-yl)methyl)-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-8-yl)piperidin-3-yl)acetamide
IUPAC Name N-[(3R)-1-[7-but-2-ynyl-3-methyl-1-[(4-methylquinazolin-2-yl)methyl]-2,6-dioxopurin-8-yl]piperidin-3-yl]acetamide
SMILES CC#CCn1c(nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c12)N5CCC[C@H](C5)NC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C27H30N8O3/c1-5-6-14-34-23-24(31-26(34)33-13-9-10-19(15-33)29-18(3)36)32(4)27(38)35(25(23)37)16-22-28-17(2)20-11-7-8-12-21(20)30-22/h7-8,11-12,19H,9-10,13-16H2,1-4H3,(H,29,36)/t19-/m1/s1
Formula Weight 514.2441
Percent Purity >95%
Chemical Name or Material Linagliptin Acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.