Learn More
LIME1 Mouse anti-Human, Clone: LIME-10, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human LIME1.
Brand: Abnova MAB12944.50ug
Description
LIME1 is a raft-associated transmembrane adaptor phosphoprotein that is preferentially expressed in hemopoietic cells, particularly T cells (Brdickova et al., 2003 [PubMed 14610046]; Hur et al., 2003 [PubMed 14610044]).[supplied by OMIM]
Specifications
| LIME1 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| FLJ20406/LIME/LP8067/dJ583P15.4 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 54923 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
| Immunohistochemistry (PFA fixed), Western Blot | |
| LIME-10 | |
| Mouse monoclonal antibody raised against synthetic peptide of human LIME1. | |
| In PBS, pH 7.4 (0.09% sodium azide). | |
| LIME1 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 281-296 at C-terminus region of human LIME1. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.