missing translation for 'onlineSavingsMsg'
Learn More

Abcam Licarbazepine, Antiepileptic Agent, >99% Purity, Solid, 50 mg

Product Code. 32109078
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109078 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109078 Supplier Abcam Supplier No. ab14586750mg
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antiepileptic agent and active metabolite of oxcarbazepine with >99% purity.

  • Molecular Weight: 254.28 Da
  • Purity: >99%
  • Form: Solid
  • Activity: Active metabolite of oxcarbazepine (ab141827). Induces microsomal enzymes via sodium channel protein inhibition. Potentiates minor GABAA receptor currents; exhibits anticonvulsant activity. Antiepileptic agent. Active in vivo.
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice
  • CAS Number: 29331-92-8

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 10,11-dihydro-10-hydroxy carbamazepine
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Sodium Channel Protein, GABAA Receptor
Formulation 100% 10,11-Dihydro-10-hydroxy Carbamazepine
Quantity 50 mg
For Use With (Application) Anticonvulsant research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.