missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX2 Antibody (CL6137) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61435PE
This item is not returnable.
View return policy
Description
LHX2 Monoclonal antibody specifically detects LHX2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| LHX2 | |
| Monoclonal | |
| PE | |
| hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2 | |
| This antibody was developed using a synthetic peptide derived from P50458, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Neuroscience, Stem Cells | |
| 9355 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL6137 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction