missing translation for 'onlineSavingsMsg'
Learn More

Lgi1 Antibody (S283-7), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42190

 View more versions of this product

Product Code. 18666836

  • 567.00€ / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lgi1 Monoclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lgi1
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Q9JIA1.1
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunofluorescence
S283-7
Unconjugated
PBS (pH 7.4), 50% Glycerol
epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1
Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∽50% identity with LGI2, LGI3 and LGI4.
0.1 mg
Neuroscience
9211
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.