missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€
Specifications
| Antigen | Lgi1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Lgi1 Polyclonal specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| Lgi1 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| LGI1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAQ89244 | |
| 9211 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human LGI1. The immunogen is located within the first 50 amino acids of LGI1. Amino Acid Squence: LLLTEGKKPAKPK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title