missing translation for 'onlineSavingsMsg'
Learn More

LGALS3BP Mouse anti-Human, CoraLite 488, Clone: 2D8E11, Proteintech

Mouse Monoclonal Antibody

145.00€ - 450.00€

Specifications

Antigen LGALS3BP
Clone 2D8E11
Concentration 2000 μg/mL
Applications Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17721279
View Documents
Proteintech
CL488-60066-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17711279
View Documents
Proteintech
CL488-60066-100UL
100 μL
450.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LGALS3BP Monoclonal antibody specifically detects LGALS3BP in Human samples. It is validated for IF
Specifications

Specifications

LGALS3BP
2000 μg/mL
Monoclonal
Liquid
Human
3959
Primary
Store at -20°C
2D8E11
Immunofluorescence
CoraLite 488
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
IgG2b
Protein A purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.