missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y Antibody (MB 311), DyLight 680, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28040FR
This item is not returnable.
View return policy
Description
Lewis Y Monoclonal antibody specifically detects Lewis Y in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Lewis Y | |
| Monoclonal | |
| DyLight 680 | |
| IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 | |
| Lewis Y | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| MB 311 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction