missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levonorgestrel, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Levonorgestrel
Specifications
Specifications
| Chemical Name or Material | Levonorgestrel |
| CAS | 797-63-7 |
| Analyte or Component Names | Levonorgestrel |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Levonorgestrel, (-)-Norgestrel, 18-Methylnorethindrone, 19-Nortestosterone, 17-ethynyl-18-methyl-, D-(-)-Norgestrel, D-Norgestrel, Dexnorgestrel, Femilis, Femilis Slim, Follistrel, Jadelle, Levonova, Lng-IUS, Microlut, Microluton, Microval, Mirena, Norgeston, Norgestrelem, Norlevo, Norplant, Norplant 2, Norplant II, Postinor 2, Wy 5104, (17alpha)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one, 13-Ethyl-17-ethynyl-17beta-hydroxy-4-gonen-3-one, 13-Ethyl-17alpha-ethynyl-17-hydroxygon-4-en-3-one, 13-Ethyl-17alpha-ethynylgon-4-en-17beta-ol-3-one, 13beta-Ethyl-17alpha-ethynyl-17beta-hydroxygon-4-en-3-one, 17alpha-Ethynyl-13-ethyl-19-nortestosterone, 17alpha-Ethynyl-13beta-ethyl-3-oxo-4-estren-17beta-ol, 17alpha-Ethynyl-17-hydroxy-18-methylestr-4-en-3-one, 17alpha-Ethynyl-18-homo-19-nortestosterone, 17alpha-Ethynyl-18-methyl-19-nortestosterone, 17beta-Hydroxy-18-methyl-19-nor-17alpha-pregn-4-en-20-yn-3-one, 18-Methyl-17alpha-ethynyl-19-nortestosterone, d-(17alpha)-17-Hydroxy-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one, Norgestimate Impurity B |
| InChI Formula | InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1 |
| SMILES | CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction