Learn More
(±)-Levomepromazine-d6 maleate, Honeywell Fluka™
Product Code. 15929781 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15929781
Brand: Honeywell Fluka R154310MG
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
Physical Form Solid
Industry Type Tranquilizer
Quantity 10 mg
Grade Analytical Standard