Learn More
Levodopa Methyl Ester Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Levodopa Methyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | L-DOPA Methyl Ester Hydrochloride |
| CAS | 1421-65-4 |
| Analyte or Component Names | L-DOPA Methyl Ester Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C10 H13 N O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (1:1), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride (6CI,7CI), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride, L- (8CI), L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (9CI), 3,4-Dihydroxyphenyl-L-alanine methyl ester hydrochloride, L-DOPA Me ester hydrochloride, L-Dopa methyl ester hydrochloride, Levodopa methyl ester hydrochloride, Methyl L-3-(3,4-dihydroxyphenyl)alaninate hydrochloride, ST 41769 |
| InChI Formula | InChI=1S/C10H13NO4.ClH/c1-15-10(14)7(11)4-6-2-3-8(12)9(13)5-6;/h2-3,5,7,12-13H,4,11H2,1H3;1H/t7-;/m0./s1 |
| SMILES | Cl.COC(=O)[C@@H](N)Cc1ccc(O)c(O)c1 |
| IUPAC Name | methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride |
| Show More |