Learn More
[Leu5]-Enkephalin Acetate, TRC
Product Code. 30012119
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012119 10 mg 10mg
30008918 100 mg 100mg
30011016 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012119 Supplier Toronto Research Chemicals Supplier No. TRCL33034010MG
 Restricted Item
Estimated Shipment: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81678-16-2
Synonym 1-5-Dynorphin∣L-Tyrosylglycylglycyl-L-phenylalanyl-L-leucine∣5-L-Leucine-enkephalin∣Leu-enkephalin∣Leucine Enkephalin∣Leucyl-enkephalin∣Tyr-Gly-Gly-Phe-Leu-OH
Specifications
Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 615.67
Quantity 10 mg
Shipping Condition Room Temperature