CAS RN 81678-16-2
IUPAC Name:
2-[2-(2-{2-[2-amino-3-(4-hydroxyphenyl)propanamido]acetamido}acetamido)-3-phenylpropanamido]-4-methylpentanoic acid; acetic acid
Synonyms:
acetic acid; tyr-gly-gly-phe-leu
Molecular Weight (g/mol):
615.68
Molecular Formula:
C30H41N5O9
InChi Key:
BAPJLGTVLSUVBN-UHFFFAOYNA-N
SMILES:
CC(O)=O.CC(C)CC(NC(=O)C(CC1=CC=CC=C1)NC(=O)CNC(=O)CNC(=O)C(N)CC1=CC=C(O)C=C1)C(O)=O
1
–
2
of
2
results
2
[Leu5]-Enkephalin Acetate, TRC
CAS: 81678-16-2 Synonym: 1-5-Dynorphin∣L-Tyrosylglycylglycyl-L-phenylalanyl-L-leucine∣5-L-Leucine-enkephalin∣Leu-enkephalin∣Leucine Enkephalin∣Leucyl-enkephalin∣Tyr-Gly-Gly-Phe-Leu-OH