missing translation for 'onlineSavingsMsg'
Learn More

[Leu5]-Enkephalin Acetate, TRC

Product Code. 30011016
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011016 25 mg 25mg
30012119 10 mg 10mg
30008918 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011016 Supplier Toronto Research Chemicals Supplier No. TRCL33034025MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81678-16-2
Synonym 1-5-Dynorphin∣L-Tyrosylglycylglycyl-L-phenylalanyl-L-leucine∣5-L-Leucine-enkephalin∣Leu-enkephalin∣Leucine Enkephalin∣Leucyl-enkephalin∣Tyr-Gly-Gly-Phe-Leu-OH

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Weight (g/mol) 615.67
Quantity 25 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.