missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin/OB Mouse anti-Human, Clone: 11, Novus Biologicals™
Mouse Monoclonal Antibody
497.00€
Specifications
| Antigen | Leptin/OB |
|---|---|
| Clone | 11 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Leptin/OB Monoclonal specifically detects Leptin/OB in Human samples. It is validated for Western Blot, ELISA.Specifications
| Leptin/OB | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| 3952 | |
| IgG1 κ | |
| Protein A or G purified |
| 11 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS | |
| Recombinant human leptin adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title