missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPREL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06245-50ul
This item is not returnable.
View return policy
Description
LEPREL1 Polyclonal antibody specifically detects LEPREL1 in Human samples. It is validated for Western Blot
Specifications
| LEPREL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ10718, leprecan-like 1, leprecan-like protein 1, MCVD, myxoid liposarcoma associated protein 4, myxoid liposarcoma-associated protein 4, P3H2, prolyl 3-hydroxylase 2, prolyl 3-hydroxylase 3 | |
| Produced in rabbits immunized with E. coli-derived Human LEPREL1 fragment. | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 55214 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction