missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEO1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79777CL1
This item is not returnable.
View return policy
Description
LEO1 Polyclonal antibody specifically detects LEO1 in Human,Mouse samples. It is validated for Western Blot
Specifications
| LEO1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), RDLReplicative senescence down-regulated leo1-like protein, RNA polymerase-associated protein LEO1 | |
| The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human senescence down regulated leo1-like using the numbering given in TrEMBL entry Q8WVC0 (GeneID 123169). | |
| 0.1 mL | |
| Phospho Specific, Transcription Factors and Regulators | |
| 123169 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction