missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Pneumophila Antibody (2F10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2520AF750
This item is not returnable.
View return policy
Description
Legionella Pneumophila Monoclonal antibody specifically detects Legionella Pneumophila in Bacteria samples. It is validated for ELISA
Specifications
| Legionella Pneumophila | |
| Monoclonal | |
| Alexa Fluor 750 | |
| L. pneumophila, Legionellosis | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with membrane fraction of Legionella pneumophila, strain Philadelphia 1. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 |
| ELISA | |
| 2F10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction