Learn More
Buerkle™ LDPE Round Wide Mouth Bottles
Product Code. p-8002304 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2000 mL
Unit Size:
Each
Product Code. 11856393
Brand: Buerkle™ 03180050
Only null left
Add to basket
Add to basket
Buerkle™ LDPE Round Wide Mouth Bottles with wide-necked opening and screw cap for viscous media, powder, granules and pastes.
  • Sturdy bottle with wide-necked opening and screw cap
  • Particularly suitable for viscous media, powder, granules and pastes
  • Leakproof
  • LDPE
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Color Transparent
Diameter (Metric) Inner Neck 24 mm
For Use With (Application) Suitable For Use With Foodstuff
Material LDPE
Quantity Each
Transparency Yes
Thread Size (Metric) 32 mm
Closure Color Transparent
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 80 mm
Product Type Wide Mouth Bottle
Closure Material LDPE
Diameter (Metric) 40 mm
Closure Type Screw Cap
Through the Closure Dispensing No