Learn More
BRAND™ LDPE Narrow Neck Bottle for Schilling Burettes
Shop All Brand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Each
Description
- Low-density Polyethylene (LDPE) construction
- Narrow mouth resevoir bottle
- With tube bushing
- For use with Schilling burettes
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | LDPE |
| Product Type | Reservoir Bottle |