Learn More
Buerkle™ LDPE Brown Round Wide Mouth Bottles
Product Code. p-8002298 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
Individual customer-specific printing possible - please ask us!
Product Code. 10378642
Brand: Buerkle™ 03200250
Only null left
Add to basket
Add to basket
Individual customer-specific printing possible - please ask us!
Sturdy bottle with wide-necked opening and screw cap. Particularly suitable for viscous media, powder, granules and pastes. Leakproof
  • LDPE
  • LDPE coloured brown for light-sensitive liquids
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Color Brown
Diameter (Metric) Inner Neck 31 mm
Material LDPE
Quantity Each
Thread Size (Metric) 40 mm
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 127 mm
Diameter (Metric) 64 mm
Closure Type Screw Cap