Learn More
MP Biomedicals™ LB Medium, Large Capsules
Shop All MP Biomedical Products
Click to view available options
Quantity:
227 g
454 g
1000 g
5000 g
15 kg
Unit Size:
15kg
1kg
227g
454g
5kg
Description
- Formulations for the E. coli growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Format | Capsule |
| Product Type | Microbiology Media |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L |
| For Use With (Application) | For growth of bacteria |
| Quantity | 1000 g |